Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Solvent Oil, Reagent for volumetric two-component KF titration in oils (working medium), Solstice
BIOPHARMA

Catalog No. 6000669 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-69 1 L
60-046-421 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-006-69 Supplier Solstice Supplier No. 347496X1L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium for volumetric two-component Karl Fischer titration in non-polar substances, fats and oils (methanol-hexanol-based)

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solvent Oil
CAS 288-32-4, 67-56-1, 111-27-3
Density 0.83g/mL
Quantity 1 L
Flash Point 17°C (62.6°F)
MDL Number MFCD00147197
Packaging Glass bottle
UN Number UN1993BE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.