Learn More
HYDRANAL™ - Titrant 2 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Chemical Name or Material | HYDRANAL™-Titrant 2 E |
| Density | 0.83g/mL |
| Quantity | 1 L |
| Flash Point | 12°C (53.6°F) |
| UN Number | UN1170 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.