Learn More
HYDRANAL™ - Xylene, Solvent for KF titration, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
Specifications
| CAS | 95-47-6 |
| Quantity | 1 L |
| Flash Point | 31°C (87.8°F) |
| Packaging | Glass bottle |
| UN Number | UN1307 |
| Chemical Name or Material | HYDRANAL™-Xylene |
Safety and Handling
| Hazard Statement | P210-P261-P280-P301 + P310-P305 + P351 + P338-P370 + P378 |
| RTECSNumber | ZE2450000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.