Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Xylene, Solvent for KF titration, Solstice

Catalog No. 60046672 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-672 6 x 1 L
60-006-85 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-672 Supplier Solstice Supplier No. 378666X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solvent for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym 1,2-Dimethylbenzene
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

CAS 95-47-6
CAS Min % ≥90.0000%
Boiling Point 143°C to 145°C
Density 0.88 g/cm3
Quantity 6 x 1 L
CAS Max % ≤100.0000%
Flash Point 30°C
Packaging Glass Bottle
UN Number UN1307
Formula Weight 106.17
Grade Karl Fischer
Chemical Name or Material HYDRANAL™ - Xylene, Solvent for KF titration
Show More Show Less
Hazard Statement P210-P261-P280-P301 + P310-P305 + P351 + P338-P370 + P378
ShelfLife 1800 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Xylenes
EINECSNumber 202-422-2
RTECSNumber 202-422-2
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.