Learn More
Medchemexpress LLC Hydrangenol | 480-47-7 | 1 MG
SDP
Catalog No. 5000419321
Encompass_Preferred
Hydrangenol is an orally active antiphotoaging compound isolated from Hydrangea serrata leaves. It prevents wrinkle formation by reducing MMP and inflammatory cytokine expression, and by increasing moisturizing factors and antioxidant gene levels.
  • Reduces wrinkle formation, dorsal thickness, dehydration, and collagen degradation
  • Increases expression of involucrin, filaggrin, and aquaporin-3 (AQP3)
  • Promotes hyaluronic acid (HA) production
  • Decreases matrix metalloproteinase (MMP)-1/-3, cyclooxygenase-2 (COX-2), and interleukin-6 (IL-6) expression
  • Attenuates phosphorylation of MAPKs (ERK and p38)
  • Upregulates Nrf2, HO-1, NQO-1, GCLM, and GCLC expression
Catalog No. 50-004-19321 Supplier Medchemexpress LLC Supplier No. HYN40281MG
Hydrangenol is an orally active antiphotoaging compound isolated from Hydrangea serrata leaves. It prevents wrinkle formation by reducing MMP and inflammatory cytokine expression, and by increasing moisturizing factors and antioxidant gene levels.
  • Reduces wrinkle formation, dorsal thickness, dehydration, and collagen degradation
  • Increases expression of involucrin, filaggrin, and aquaporin-3 (AQP3)
  • Promotes hyaluronic acid (HA) production
  • Decreases matrix metalloproteinase (MMP)-1/-3, cyclooxygenase-2 (COX-2), and interleukin-6 (IL-6) expression
  • Attenuates phosphorylation of MAPKs (ERK and p38)
  • Upregulates Nrf2, HO-1, NQO-1, GCLM, and GCLC expression