Learn More
TARGETMOL CHEMICALS INC HYDRANGENOL 5MG
SDP
Catalog No. 502578872
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hydrangenol is a natural product and can be isolated from Hydrangea serrata leaves. Hydrangenol is an antiphotoaging compound with oral activity. Hydrangenol can reduce MMP and inflammatory cytokine expression and increase moisturizing factors and antioxidant genes level result in preventing wrinkle formation. purity: 99%
Catalog No. 50-257-8872 Supplier TARGETMOL CHEMICALS INC Supplier No. TN42345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hydrangenol is a natural product and can be isolated from Hydrangea serrata leaves. Hydrangenol is an antiphotoaging compound with oral activity. Hydrangenol can reduce MMP and inflammatory cytokine expression and increase moisturizing factors and antioxidant genes level result in preventing wrinkle formation. purity: 99%