Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrastinine | 6592-85-4 | 99.92% | 5 MG
SDP

Catalog No. 5000412852
Encompass_Preferred

Hydrastinine is a major alkaloid constituent in goldenseal (Hydrastis canadensis) and can be used as a haemostatic agent. Goldenseal has been used for the treatment of various ailments including gastrointestinal disturbances, urinary tract disorders, and inflammation. The five major alkaloid constituents in goldenseal are Berberine, Palmatine, Hydrastine, Hydrastinine, and Canadine.

  • Molecular weight: 207.23
  • Formula: C11H13NO3
  • Appearance: Solid
  • Color: Off-white to light brown
  • Structure classification: Alkaloids, Isoquinoline Alkaloids
  • Initial source: Plants, Ranunculaceae

Catalog No. 50-004-12852 Supplier Medchemexpress LLC Supplier No. HYB1181A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrastinine is a major alkaloid constituent in goldenseal (Hydrastis canadensis) and can be used as a haemostatic agent. Goldenseal has been used for the treatment of various ailments including gastrointestinal disturbances, urinary tract disorders, and inflammation. The five major alkaloid constituents in goldenseal are Berberine, Palmatine, Hydrastine, Hydrastinine, and Canadine.

  • Molecular weight: 207.23
  • Formula: C11H13NO3
  • Appearance: Solid
  • Color: Off-white to light brown
  • Structure classification: Alkaloids, Isoquinoline Alkaloids
  • Initial source: Plants, Ranunculaceae

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.