Learn More
Medchemexpress LLC Hydrastinine hydrochloride | 4884-68-8 | MFCD09763656 | 99.4% | 243.69 g·mol⁻1 | C11H14ClNO3 | 10 MG
SDP
Catalog No. 5000298144
Encompass_Preferred
Hydrastinine hydrochloride is the hydrochloride salt of hydrastinine, an alkaloid found in goldenseal (Hydrastis canadensis). It is supplied for research and analytical use and is reported to have haemostatic activity. The product is offered in solid form and as DMSO solutions, with purity levels suitable for analytical and pharmacological studies.
  • High purity (~99.4%), suitable for analytical applications.
  • Available as solid and as 10 mM solutions in DMSO.
  • Reported haemostatic activity used in pharmacology research.
  • Supplied for research and analytical purposes only, not for human use.
  • Key identifiers: CAS 4884-68-8, formula C11H14ClNO3, molecular weight 243.69 g·mol⁻1.
Catalog No. 50-002-98144 Supplier Medchemexpress LLC Supplier No. HYB118110MG
Hydrastinine hydrochloride is the hydrochloride salt of hydrastinine, an alkaloid found in goldenseal (Hydrastis canadensis). It is supplied for research and analytical use and is reported to have haemostatic activity. The product is offered in solid form and as DMSO solutions, with purity levels suitable for analytical and pharmacological studies.
  • High purity (~99.4%), suitable for analytical applications.
  • Available as solid and as 10 mM solutions in DMSO.
  • Reported haemostatic activity used in pharmacology research.
  • Supplied for research and analytical purposes only, not for human use.
  • Key identifiers: CAS 4884-68-8, formula C11H14ClNO3, molecular weight 243.69 g·mol⁻1.