Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whirl-Pak™ Hydrated PolyProbe™ Sampling Bag, Sponge with Stick
GREENER_CHOICESDP

Catalog No. 14209183 Shop All Whirl-Pak Products
Change view
Click to view available options
Capacity (Metric):
710 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-209-183 710 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-209-183 Supplier Whirl-Pak™ Supplier No. B01592
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydrated PolyProbe is designed for surface sampling and effective in hard-to-reach areas.

  • Durable polyurethane sponge that resists tearing and fraying
  • 8 in. polypropylene holder
  • Packaged in a sealed 24 oz. (710 mL) write-on bag
  • Hydrated with 10 mL of HiCap™ Neutralizing Broth

Specifications

Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
Material Polyethylene
Sterility Sterile
Fill Volume 24 oz.
Latex Free Yes
No. per Pack 100/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Foil Pouch
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 3 mil
Dimensions (L x W) 6 x 9 in. (15 x 23 cm)
Length (English) 9 in.
Width (English) 6 in.
Product Type Sample Bag with Sampling Sponge
Capacity (English) 24 oz.
Capacity (Metric) 710 mL
Length (Metric) 23 cm
Width (Metric) 15 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.