Learn More
MSC Enerpac Hydraulic Bottle Jack, 3 Tons, 6.625 in. to 13.31 in., 1 ea.
Catalog No. 5000516063
Encompass
This hydraulic bottle jack is a durable lifting tool constructed from high-strength steel, designed for reliable performance in demanding industrial and maintenance applications. It features a 3-ton load capacity and offers a versatile height range to facilitate various lifting tasks.
  • Features a heat-treated, adjustable extension screw with a cleated saddle to improve adjustment precision and prevent load slipping
  • Equipped with a safety relief valve to prevent overloading and an automatic by-pass port to prevent over-extension
  • Designed with a thick base and large surface area to provide increased stability and strength
  • Includes high-strength beam and pump linkage for extended operational life
  • Features a wiper seal to protect against contaminants
  • Fully serviceable design with a dedicated pumping handle
  • Meets or exceeds ANSI, PALD, and CE standards
Catalog No. 50-005-16063 Supplier MSC Supplier No. 96106992
This hydraulic bottle jack is a durable lifting tool constructed from high-strength steel, designed for reliable performance in demanding industrial and maintenance applications. It features a 3-ton load capacity and offers a versatile height range to facilitate various lifting tasks.
  • Features a heat-treated, adjustable extension screw with a cleated saddle to improve adjustment precision and prevent load slipping
  • Equipped with a safety relief valve to prevent overloading and an automatic by-pass port to prevent over-extension
  • Designed with a thick base and large surface area to provide increased stability and strength
  • Includes high-strength beam and pump linkage for extended operational life
  • Features a wiper seal to protect against contaminants
  • Fully serviceable design with a dedicated pumping handle
  • Meets or exceeds ANSI, PALD, and CE standards