Learn More
Grainger Hydraulic Hose Cap 1-5/8 -12 Female JIC
Catalog No. 502749796
Encompass_Preferred
The Aeroquip Hydraulic Hose Cap is a steel hydraulic hose cap with a 1/4" NPTF thread size. It is made of carbon steel and has a maximum pressure rating of 5000 psi. The cap is designed to protect hydraulic hose ends and prevent contamination. It measures 0.81" in diameter and 0.5" in height. This reusable cap screws onto exposed hydraulic hose fittings to seal hose ends from dirt and moisture when disconnected. It provides an economical way to cap off hydraulic hoses.
Catalog No. 50-274-9796 Supplier Grainger Supplier No. 14L784
The Aeroquip Hydraulic Hose Cap is a steel hydraulic hose cap with a 1/4" NPTF thread size. It is made of carbon steel and has a maximum pressure rating of 5000 psi. The cap is designed to protect hydraulic hose ends and prevent contamination. It measures 0.81" in diameter and 0.5" in height. This reusable cap screws onto exposed hydraulic hose fittings to seal hose ends from dirt and moisture when disconnected. It provides an economical way to cap off hydraulic hoses.