Learn More
Grainger Hydraulic Hose Cap 2-1/2 -12 Female JIC
Catalog No. 502749748
Encompass_Preferred
The Aeroquip Hydraulic Hose Cap is a steel hydraulic hose cap with a 1/4" NPTF thread size. It is made of steel for durability and has a maximum pressure rating of 5000 psi. This hydraulic hose cap is designed to protect hydraulic hose ends and prevent leaks. It measures 0.81" in diameter and 0.5" in height. The Aeroquip hydraulic hose cap comes in a pack of 10 and can be used with Aeroquip hoses and fittings. It provides an economical way to cap off hydraulic hoses. This steel hydraulic hose cap is made in the USA.
Catalog No. 50-274-9748 Supplier Grainger Supplier No. 14L786
The Aeroquip Hydraulic Hose Cap is a steel hydraulic hose cap with a 1/4" NPTF thread size. It is made of steel for durability and has a maximum pressure rating of 5000 psi. This hydraulic hose cap is designed to protect hydraulic hose ends and prevent leaks. It measures 0.81" in diameter and 0.5" in height. The Aeroquip hydraulic hose cap comes in a pack of 10 and can be used with Aeroquip hoses and fittings. It provides an economical way to cap off hydraulic hoses. This steel hydraulic hose cap is made in the USA.