Learn More
Grainger Hydraulic Impact Wrench 7.4 lb
Catalog No. 502766364
Encompass_Preferred
The Grainger 16V989 is a heavy duty, high capacity, industrial grade battery charger. It features a fully automatic 8 amp charging system to charge 12V lead acid batteries from 32-300Ah. The charger has adjustable voltage settings for optimal charging and maintenance of flooded, gel, and AGM type batteries. It includes overcharge protection, reverse polarity protection, and automatic shutoff when the battery is fully charged. The durable steel housing allows for indoor or outdoor use. This is an ideal battery charger for warehouses, manufacturing facilities, construction sites, and other commercial/industrial applications.
Catalog No. 50-276-6364 Supplier Grainger Supplier No. 16V989
The Grainger 16V989 is a heavy duty, high capacity, industrial grade battery charger. It features a fully automatic 8 amp charging system to charge 12V lead acid batteries from 32-300Ah. The charger has adjustable voltage settings for optimal charging and maintenance of flooded, gel, and AGM type batteries. It includes overcharge protection, reverse polarity protection, and automatic shutoff when the battery is fully charged. The durable steel housing allows for indoor or outdoor use. This is an ideal battery charger for warehouses, manufacturing facilities, construction sites, and other commercial/industrial applications.