Learn More
Grainger Hydraulic Lift Kt 4 Post Elctrc 1000 lb
Catalog No. 502766864
Encompass_Preferred
The Grainger 4-Post Electric Hydraulic Lift Table has a lifting capacity of 1,000 lbs and a lowered height of 24". It features an electric hydraulic lift system with push-button controls for smooth, precise lifting. The lift table has a rugged steel construction with a powder coat finish and four swivel casters for easy mobility. It can lift items up to 30" high for improved ergonomics and material handling. This versatile lift table is ETL certified to meet North American safety standards. With its 1,000 lb capacity and electric operation, this Grainger lift table is ideal for warehouses, distribution centers, and manufacturing facilities.
Catalog No. 50-276-6864 Supplier Grainger Supplier No. 16Y679
The Grainger 4-Post Electric Hydraulic Lift Table has a lifting capacity of 1,000 lbs and a lowered height of 24". It features an electric hydraulic lift system with push-button controls for smooth, precise lifting. The lift table has a rugged steel construction with a powder coat finish and four swivel casters for easy mobility. It can lift items up to 30" high for improved ergonomics and material handling. This versatile lift table is ETL certified to meet North American safety standards. With its 1,000 lb capacity and electric operation, this Grainger lift table is ideal for warehouses, distribution centers, and manufacturing facilities.