Learn More
Grainger Hydraulic Lift Kt 4 Post Elctrc 1500 lb
Catalog No. 502767605
Encompass_Preferred
The Grainger Hydraulic Lift Kit is a 4-post electric hydraulic lift with a lifting capacity of 1500 lbs. It has a lifting height range of 39"-70" and comes with everything needed for installation including columns, adapters, hoses, fittings, and hardware. The lift features heavy duty welded steel construction with a durable powder coat finish. It can lift most passenger vehicles for service and maintenance and includes an electric hydraulic power unit that plugs into a standard wall outlet. Safety features include hydraulic velocity fuses and mechanical locks. Overall this is a high quality 4-post lift kit for home or professional garage use.
Catalog No. 50-276-7605 Supplier Grainger Supplier No. 16Y696
The Grainger Hydraulic Lift Kit is a 4-post electric hydraulic lift with a lifting capacity of 1500 lbs. It has a lifting height range of 39"-70" and comes with everything needed for installation including columns, adapters, hoses, fittings, and hardware. The lift features heavy duty welded steel construction with a durable powder coat finish. It can lift most passenger vehicles for service and maintenance and includes an electric hydraulic power unit that plugs into a standard wall outlet. Safety features include hydraulic velocity fuses and mechanical locks. Overall this is a high quality 4-post lift kit for home or professional garage use.