Learn More
Grainger Hydraulic Lift Kt 4 Post Elctrc 1000 lb
Catalog No. 502767880
Encompass_Preferred
The Grainger 4-Post Electric Hydraulic Lift Table has a lifting capacity of 1,000 lbs. It features a hydraulic lifting system to raise and lower the table top from 24" to 36" high. The lift table has a 48" x 48" steel top and is constructed of welded steel with a powder coat finish. It comes with non-marring 3" casters, two of which lock. The lift is operated by an electric hydraulic pump with an integrated control panel and has a lift speed of 2"/second. This durable lift table is ideal for material handling and work positioning needs.
Catalog No. 50-276-7880 Supplier Grainger Supplier No. 16Y677
The Grainger 4-Post Electric Hydraulic Lift Table has a lifting capacity of 1,000 lbs. It features a hydraulic lifting system to raise and lower the table top from 24" to 36" high. The lift table has a 48" x 48" steel top and is constructed of welded steel with a powder coat finish. It comes with non-marring 3" casters, two of which lock. The lift is operated by an electric hydraulic pump with an integrated control panel and has a lift speed of 2"/second. This durable lift table is ideal for material handling and work positioning needs.