Learn More
Grainger Hydraulic Lift Kt 4 Post Elctrc 1000 lb
Catalog No. 502767823
Encompass_Preferred
The 4-Post Hydraulic Lift Table from Grainger is an electric hydraulic lift table with a 1000 lb capacity. It has a 48" x 48" platform and a lifting range of 23"-33". The lift table is constructed of steel with a powder coat finish and features dual hydraulic cylinders, an electric hydraulic pump, and push button controls. Safety features include mechanical locks and an anti-burst safety valve. This heavy duty lift assists with material handling and height adjustment in industrial environments.
Catalog No. 50-276-7823 Supplier Grainger Supplier No. 16Y678
The 4-Post Hydraulic Lift Table from Grainger is an electric hydraulic lift table with a 1000 lb capacity. It has a 48" x 48" platform and a lifting range of 23"-33". The lift table is constructed of steel with a powder coat finish and features dual hydraulic cylinders, an electric hydraulic pump, and push button controls. Safety features include mechanical locks and an anti-burst safety valve. This heavy duty lift assists with material handling and height adjustment in industrial environments.