Learn More
Grainger Hydraulic Lift Kt 4 Post Elctrc 1500 lb
Catalog No. 502768117
Encompass_Preferred
The Dyna-Lift Hydraulic Lift Kit is a 10 inch stroke hydraulic lift kit designed for lifting and lowering heavy loads. It has a lifting capacity of 5,000 lbs and comes with a hydraulic cylinder, hoses, fittings, pump, and reservoir. The cylinder has a 2 inch bore and 1.5 inch rod. The pump operates at 700 psi and has a flow rate of 0.75 gpm. The kit is made in the USA out of high quality components for industrial lifting applications.
Catalog No. 50-276-8117 Supplier Grainger Supplier No. 16Y697
The Dyna-Lift Hydraulic Lift Kit is a 10 inch stroke hydraulic lift kit designed for lifting and lowering heavy loads. It has a lifting capacity of 5,000 lbs and comes with a hydraulic cylinder, hoses, fittings, pump, and reservoir. The cylinder has a 2 inch bore and 1.5 inch rod. The pump operates at 700 psi and has a flow rate of 0.75 gpm. The kit is made in the USA out of high quality components for industrial lifting applications.