Learn More
Grainger Hydraulic Lift Kt 4 Post Mnl 1000 lb
Catalog No. 502766273
Encompass_Preferred
The Dyna-Lift Hydraulic Lift Kit Workbench is a heavy-duty hydraulic lift table that raises and lowers to provide an adjustable work surface height from 30" to 48". It has a weight capacity of 1,000 lbs and a 48" x 24" steel top worktable. The hydraulic lift mechanism provides smooth and precise height adjustments. It also features industrial casters and dual locking brakes for easy mobility and stability. The steel construction ensures durability, and the powder coat finish resists wear. This versatile workbench allows working at the most ergonomic height.
Catalog No. 50-276-6273 Supplier Grainger Supplier No. 16Y672
The Dyna-Lift Hydraulic Lift Kit Workbench is a heavy-duty hydraulic lift table that raises and lowers to provide an adjustable work surface height from 30" to 48". It has a weight capacity of 1,000 lbs and a 48" x 24" steel top worktable. The hydraulic lift mechanism provides smooth and precise height adjustments. It also features industrial casters and dual locking brakes for easy mobility and stability. The steel construction ensures durability, and the powder coat finish resists wear. This versatile workbench allows working at the most ergonomic height.