Learn More
Grainger Hydraulic Lift Kt 4 Post Mnl 1000 lb
Catalog No. 502766276
Encompass_Preferred
The Dyna-Lift Hydraulic Lift Kit Workbench is a heavy-duty workbench with a hydraulic lift system that allows the work surface to be raised and lowered smoothly and easily with the turn of a crank. The workbench has a steel frame with a laminated maple work surface that measures 60"W x 24"D. The work surface can be raised from 30" up to 42" high to accommodate working while standing or sitting. The lift capacity is 300 lbs with an adjustable height range of 12". This professional-grade workbench also features two steel shelves below the work surface for storage. The hydraulic lift uses a manual crank to raise and lower the work surface. Assembly is required.
Catalog No. 50-276-6276 Supplier Grainger Supplier No. 16Y670
The Dyna-Lift Hydraulic Lift Kit Workbench is a heavy-duty workbench with a hydraulic lift system that allows the work surface to be raised and lowered smoothly and easily with the turn of a crank. The workbench has a steel frame with a laminated maple work surface that measures 60"W x 24"D. The work surface can be raised from 30" up to 42" high to accommodate working while standing or sitting. The lift capacity is 300 lbs with an adjustable height range of 12". This professional-grade workbench also features two steel shelves below the work surface for storage. The hydraulic lift uses a manual crank to raise and lower the work surface. Assembly is required.