Learn More
Grainger Hydraulic Lift Kt 4 Post Mnl 1000 lb
Catalog No. 502766272
Encompass_Preferred
The Grainger Approved Hydraulic Lift Kit is a hydraulic lift kit for lifting heavy equipment. It has a lifting capacity of 10 tons (20,000 lbs) and a lift range from 14.5" to 35.5". The kit includes 2 jacks, a hydraulic power unit, and hoses. It is designed for garage service lifting and can lift most cars, trucks, tractors and other heavy equipment. The jacks feature welded steel bases and lift arms along with hydraulic cylinders for smooth lifting. This professional grade hydraulic lift kit provides portable lifting power for mechanics and auto shops.
Catalog No. 50-276-6272 Supplier Grainger Supplier No. 16Y671
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Grainger Approved Hydraulic Lift Kit is a hydraulic lift kit for lifting heavy equipment. It has a lifting capacity of 10 tons (20,000 lbs) and a lift range from 14.5" to 35.5". The kit includes 2 jacks, a hydraulic power unit, and hoses. It is designed for garage service lifting and can lift most cars, trucks, tractors and other heavy equipment. The jacks feature welded steel bases and lift arms along with hydraulic cylinders for smooth lifting. This professional grade hydraulic lift kit provides portable lifting power for mechanics and auto shops.