Learn More
Grainger Hydraulic Power Saw Replacement Guidebar
Catalog No. 502766193
Encompass_Preferred
The Grainger 16V978 is a Westward 3/8-inch drive impact socket adapter made of chrome vanadium steel with a black phosphate finish. It has a 1/2-inch square drive to accept 3/8-inch drive impact sockets. The adapter measures 2-1/2 inches long and weighs 0.19 lbs. It is rated up to 20,000 lbs of bolt breakaway torque. The adapter allows use of 3/8-inch drive impact sockets on tools with a 1/2-inch square drive.
Catalog No. 50-276-6193 Supplier Grainger Supplier No. 16V978
The Grainger 16V978 is a Westward 3/8-inch drive impact socket adapter made of chrome vanadium steel with a black phosphate finish. It has a 1/2-inch square drive to accept 3/8-inch drive impact sockets. The adapter measures 2-1/2 inches long and weighs 0.19 lbs. It is rated up to 20,000 lbs of bolt breakaway torque. The adapter allows use of 3/8-inch drive impact sockets on tools with a 1/2-inch square drive.