Learn More
Hach Company HydraVer 2 Hydrazine Reagent, 500 mL
Supplier:  Hach Company 179049
Encompass
For hydrazine determination in boiler waters by the p-Dimethylaminobenzaldehyde method. Hach method 8141. Range: 4-600 μg/L N2H4. (Also used in visual test kits. Range: 0.02-1.00 mg/L) 500 mL bottle
Catalog No. 50-199-4601