Learn More
Hach Company HydraVer 2 Hydrazine Reagent, 100 mL
Supplier:  Hach Company 179032
Encompass
For hydrazine determination in boiler waters by the p-Dimethylaminobenzaldehyde method. Hach method 8141. Range: 4 - 600 μg/L N₂H₂. (Also used in visual test kits. Range: 0.02-1.00 mg/L) 100 mL bottle.
Catalog No. 50-199-4600