Learn More
Medchemexpress LLC Hydrazide-peg4-desthiobiotin | 00-00-0 | 95.0% | 475.58 | C21H41N5O7 | 5 MG
SDP
Catalog No. 5000222477
Encompass_Preferred
Hydrazide-PEG4-Desthiobiotin is a PEG-based PROTAC linker providing a hydrazide functional group and a PEG4 spacer for conjugation in targeted protein degradation research. It is supplied at research-grade purity for use in PROTAC assembly and bioconjugation workflows.
  • Provides a hydrazide functional handle for conjugation.
  • Contains a PEG4 spacer to improve solubility and linker flexibility.
  • Suitable for PROTAC synthesis and bioconjugation applications.
  • High purity (95.0%) for consistent experimental results.
  • Store protected from light at -20 °C to maintain stability.
Catalog No. 50-002-22477 Supplier Medchemexpress LLC Supplier No. HY1347225MG
Hydrazide-PEG4-Desthiobiotin is a PEG-based PROTAC linker providing a hydrazide functional group and a PEG4 spacer for conjugation in targeted protein degradation research. It is supplied at research-grade purity for use in PROTAC assembly and bioconjugation workflows.
  • Provides a hydrazide functional handle for conjugation.
  • Contains a PEG4 spacer to improve solubility and linker flexibility.
  • Suitable for PROTAC synthesis and bioconjugation applications.
  • High purity (95.0%) for consistent experimental results.
  • Store protected from light at -20 °C to maintain stability.