Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine hydrate, 100% (Hydrazine, 64%)

Catalog No. ac196711000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196711000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196711000 Supplier Thermo Scientific Chemicals Supplier No. 196711000
Only null left
Add to Cart

CAS: 10217-52-4 | H4N2 | 32.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine;hydrate
SMILES NN

Specifications

Boiling Point 120.1°C
Melting Point -51.5°C
Color Colorless
Physical Form Liquid
Quantity 100 g
Linear Formula H2NNH2·XH2O
Flash Point 75°C
Fieser 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
Merck Index 14, 4771
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, insoluble in chloroform and ether
Formula Weight 32.04
Percent Purity 100.00 to 100.93% (N2H4.xH2O), 64.00 to 64.60% (N2H4)
Packaging Glass bottle
Refractive Index 1.4285 to 1.4315
Specific Gravity 1.0315
Density 1.0315g/mL
Chemical Name or Material Hydrazine hydrate, Hydrazine Hydrate 100% (Hydrazine, 64%)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Very toxic to aquatic life with l
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre
RTECSNumber MV8050000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.