Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Hydrate Solution, 24 to 26% (iodometric), Solstice

Catalog No. 60044909 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-909 6 x 1 L Poly Bottle
60-044-908 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-909 Supplier Solstice Supplier No. 156226X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

24-26% (iodometric)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10217-52-4 , 7732-18-5
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine;hydrate
SMILES NN

Specifications

Color Colorless
Physical Form Liquid
Quantity 6 x 1 L
Linear Formula NH2NH2 · H2O
UN Number UN3293
Beilstein 878137
Flash Point 100°C (212°F)
Formula Weight 50.06g/mol
Percent Purity 24 to 26% (iodometric)
Packaging Poly Bottle
Chemical Name or Material Hydrazine Hydrate Solution
Show More Show Less
Hazard Statement P201-P260-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P313
Precautionary Statement H302-H314-H317-H331-H350-H410
DOTInformation 6.1/PGIII
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.