Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Hydrazine Refill Kit
SDP

Catalog No. 13299639
Catalog No. 13-299-639 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R5005
Add to Cart
Add to Cart

Refill kit for the K-5005 Hydrazine visual test kit. Contains 30 self-filling ampoules for hydrazine analysis in water.

  • In an acidic solution, hydrazine reacts with PDMAB (p?dimethylaminobenzaldehyde) to form a yellow colored complex In direct proportion to the hydrazine concentration.
  • Results are expressed in ppm (mg/L) N2H4

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-5005
No. of Tests 30
Type Hydrazine Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.