Learn More
LaMotte™ Hydrazine SMART 3 Reagent System
SDPSCI_ED
Catalog No. S95004 Shop All LaMotte Company Products
Change view
Click to view available options
No. of Tests:
50
Catalog No. No. of Tests
S95004 50
1 options
Catalog No. S95004 Supplier LaMotte™ Supplier No. 365601SC
Hydrazine reagent system for use with SMART 3 Colorimeter.
  • Range 0.01 to 0.75 ppm
  • 50 Tests
  • P-Dimethylaminobenzaldehyde Method
Specifications
For Use With (Application) For Industrial Waters
For Use With (Equipment) SMART 3 Colorimeter
Material Hydrazine A
No. of Tests 50
Range 0.01 to 0.75 ppm
Test Method P-Dimethylaminobenzaldehyde Method
Type Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov