Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrazine Solution

Catalog No. 901596100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-159-6100ML 5 g
90-159-625ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 90-159-6100ML Supplier Sigma Aldrich Supplier No. 901596100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrazine promotes the cation exchange reaction for fast doping Cu ions into ZnSe nanocrystals.Valuable building block provided in ethanol for convenient handling.Hydrazine solution may be employed for the following studies:As reducing agent for the reduction of the first row (3d) transition metal complexes in aqueous solution.Preparation of nanoscale reduced metal oxides such as VO2(B), Cr2O3.As fuel for the direct hydrazine fuel cell (DHFC).As reducing agent for the synthesis of metallic monodisperse copper nanoparticles.

Specifications

CAS 657408-07-6
Melting Point 164°C to 166°C; 165.5°C
Molecular Formula C26H35O2P
Linear Formula C26H35O2P
Refractive Index n/D 1.3666
MDL Number MFCD05861611
Quantity 5 g
Synonym 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl; S Phos
Molecular Weight (g/mol) 410.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.