Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrazine solution

Catalog No. 502899303
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-9303 500 g
50-180-8930 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-289-9303 Supplier Sigma Aldrich Supplier No. 309400500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The Janus micromotors are self-propelled at a very high speed in a 0.001% hydrazine solution owing to osmotic effects.Hydrazine solution was used in the synthesis of molecular metal chalcogenide, (N2H4)3(N2H5)4Sn2Se6 complex, a precursor for SnSe2 deposition on TiO2 nanocrystalline porous films.
TRUSTED_SUSTAINABILITY

Specifications

CAS 302-01-2
Density 1.011 g/mL (at 25°C)
Molecular Formula H4N2
Linear Formula NH2NH2
Quantity 500 g
MDL Number MFCD00011417
Molecular Weight (g/mol) 32.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.