Learn More
Hach Company Hydrazine Sulfate, 100 g
Catalog No. 501994442 Shop All Hach Company Products
Encompass
ACS Grade, CAS No. 10034-93-2. H2NNH2·...H2SO4, FW = 130.12. Assay: ≥99.0% Suitable for preparation of formazin turbidity standards
Catalog No. 50-199-4442 Supplier Hach Company Supplier No. 74226
ACS Grade, CAS No. 10034-93-2. H2NNH2·...H2SO4, FW = 130.12. Assay: ≥99.0% Suitable for preparation of formazin turbidity standards