Learn More
Medchemexpress LLC Hydrazine sulfate 9 25g
SDP
Catalog No. 5000204868
Encompass_Preferred
Hydrazine sulfate is an orally active PEPCK inhibitor Hydrazine sulfate inhibits PEPCK and low-Km ALDH Hydrazine sulfate impairs gluconeogenesis and enhances the protective effect of energy substrates against MPP toxicity Hydrazine sulfate exacerbates liver damage in rats when combined with Ethanol Hydrazine sulfate can be used in the research of shock non-small cell lung cancer colorectal cancer and Parkinson s disease[1][2][3][4][5][6]
Catalog No. 50-002-04868 Supplier Medchemexpress LLC Supplier No. 25GHYW115752
Hydrazine sulfate is an orally active PEPCK inhibitor Hydrazine sulfate inhibits PEPCK and low-Km ALDH Hydrazine sulfate impairs gluconeogenesis and enhances the protective effect of energy substrates against MPP toxicity Hydrazine sulfate exacerbates liver damage in rats when combined with Ethanol Hydrazine sulfate can be used in the research of shock non-small cell lung cancer colorectal cancer and Parkinson s disease[1][2][3][4][5][6]