Learn More
Medchemexpress LLC Hydrazine sulfate 9 5kg
SDP
Catalog No. 5000206586
Encompass_Preferred
Hydrazine sulfate is an orally active PEPCK inhibitor Hydrazine sulfate inhibits PEPCK and low-Km ALDH Hydrazine sulfate impairs gluconeogenesis and enhances the protective effect of energy substrates against MPP toxicity Hydrazine sulfate exacerbates liver damage in rats when combined with Ethanol Hydrazine sulfate can be used in the research of shock non-small cell lung cancer colorectal cancer and Parkinson s disease[1][2][3][4][5][6]
Catalog No. 50-002-06586 Supplier Medchemexpress LLC Supplier No. HYW1157525KG
Hydrazine sulfate is an orally active PEPCK inhibitor Hydrazine sulfate inhibits PEPCK and low-Km ALDH Hydrazine sulfate impairs gluconeogenesis and enhances the protective effect of energy substrates against MPP toxicity Hydrazine sulfate exacerbates liver damage in rats when combined with Ethanol Hydrazine sulfate can be used in the research of shock non-small cell lung cancer colorectal cancer and Parkinson s disease[1][2][3][4][5][6]