Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydrazinecarboselenoamide | 21198-79-8 | 95.0% | 50 MG
SDP

Catalog No. 5000342887
Encompass_Preferred

Selenosemicarbazide, also known as Hydrazinecarboselenoamide, is a chemical compound with the molecular formula CH5N3Se and a molecular weight of 138.03. This off-white to gray solid has a purity of 95.0% by NMR. Derivatives of Selenosemicarbazide have shown indications of antimicrobial activity and potential anti-tumor effects.

  • Off-white to gray solid
  • Purity of 95.0%
  • Potential for antimicrobial activity
  • Potential for anti-tumor effects (derivatives)
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month

Catalog No. 50-003-42887 Supplier Medchemexpress LLC Supplier No. HY12822250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selenosemicarbazide, also known as Hydrazinecarboselenoamide, is a chemical compound with the molecular formula CH5N3Se and a molecular weight of 138.03. This off-white to gray solid has a purity of 95.0% by NMR. Derivatives of Selenosemicarbazide have shown indications of antimicrobial activity and potential anti-tumor effects.

  • Off-white to gray solid
  • Purity of 95.0%
  • Potential for antimicrobial activity
  • Potential for anti-tumor effects (derivatives)
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.