Learn More
Medchemexpress LLC Hydrazinecarbothioamide, 2-(1H-indol-3-ylmethylene)-N-(4-methylphenyl)-, (2E)- | 2097787-75-0 | 99.0% | 308.40 | 5 MG
SDP
Catalog No. 5000392238
Encompass_Preferred
Nur77 modulator 3 (HY-162147) is a chemical compound intended for research use only. It has a molecular weight of 308.40 and exhibits a high purity of 99.01% via HPLC analysis. This compound is identified by its CAS Number 2097787-75-0 and the chemical name Hydrazinecarbothioamide, 2-(1H-indol-3-ylmethylene)-N-(4-methylphenyl)-, (2E)-.
  • Provided as a white to off-white solid
  • Structure confirmed by ¹H NMR and LCMS
  • High purity of 99.01% verified by HPLC
  • Powder storage recommended at -20°C for up to 3 years
  • In-solvent storage recommended at -80°C for 6 months or -20°C for 1 month
  • Complies with specified quality standards
Catalog No. 50-003-92238 Supplier Medchemexpress LLC Supplier No. HY1621475MG
Nur77 modulator 3 (HY-162147) is a chemical compound intended for research use only. It has a molecular weight of 308.40 and exhibits a high purity of 99.01% via HPLC analysis. This compound is identified by its CAS Number 2097787-75-0 and the chemical name Hydrazinecarbothioamide, 2-(1H-indol-3-ylmethylene)-N-(4-methylphenyl)-, (2E)-.
  • Provided as a white to off-white solid
  • Structure confirmed by ¹H NMR and LCMS
  • High purity of 99.01% verified by HPLC
  • Powder storage recommended at -20°C for up to 3 years
  • In-solvent storage recommended at -80°C for 6 months or -20°C for 1 month
  • Complies with specified quality standards