Learn More
Medchemexpress LLC Hydrazinecarbothioamide, 2-(1H-indol-3-ylmethylene)-N-(4-methylphenyl)-, (2E)- | 2097787-75-0 | 99.0% | 308.40 g/mol | C17H16N4S | 10 MG
SDP
Catalog No. 5000288890
Encompass_Preferred
Nur77 modulator 3 is a small-molecule research compound that binds nuclear receptor Nur77 (NR4A1) and modulates its activity. It has been reported to inhibit TGF-β1-induced α-SMA and COLA1 expression and to reduce hepatic stellate cell activation and extracellular matrix deposition. Technical identifiers include CAS 2097787-75-0, formula C17H16N4S, molecular weight 308.40 g/mol, and purity 99.01%.
  • Modulates Nur77 activity in cellular and in vivo models.
  • Inhibits TGF-β1-induced α-SMA and COLA1 expression.
  • High purity (99.01%) suitable for biochemical and cell-based assays.
  • Supplied in small quantities appropriate for assay development and pilot studies.
Catalog No. 50-002-88890 Supplier Medchemexpress LLC Supplier No. HY16214710MG
Nur77 modulator 3 is a small-molecule research compound that binds nuclear receptor Nur77 (NR4A1) and modulates its activity. It has been reported to inhibit TGF-β1-induced α-SMA and COLA1 expression and to reduce hepatic stellate cell activation and extracellular matrix deposition. Technical identifiers include CAS 2097787-75-0, formula C17H16N4S, molecular weight 308.40 g/mol, and purity 99.01%.
  • Modulates Nur77 activity in cellular and in vivo models.
  • Inhibits TGF-β1-induced α-SMA and COLA1 expression.
  • High purity (99.01%) suitable for biochemical and cell-based assays.
  • Supplied in small quantities appropriate for assay development and pilot studies.