Learn More
Abcam 4-Hydrazino-7-nitrobenzofurazane hydrazine adduct, 25MG
Catalog No. 50996192
Encompass_Preferred
MW 227.18 Da, Purity >98%. Fluorescent derivatization reagent useful for aliphatic aldehyde detection including labeling of reducing carbohydrates.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-192 Supplier Abcam Supplier No. AB27535925MG
MW 227.18 Da, Purity >98%. Fluorescent derivatization reagent useful for aliphatic aldehyde detection including labeling of reducing carbohydrates.
The product is subject to the following: Abcam Restricted Use Statement