Learn More
Alpha Resources Inc Certified Reference Material (CRM), Titanium pin, Hydrogen 0.0046%, Oxygen 0.0453%, Nitrogen 0.0025%, Nominal Weight 0.1 g per pin, Minimum Sample Size 1 pin
SDP
Catalog No. NC2232903
Encompass
Certified Reference Material (CRM), Titanium pin, Hydrogen 0.0046%, Oxygen 0.0453%, Nitrogen 0.0025%, Nominal Weight 0.1 g per pin, Minimum Sample Size 1 pin
This item is not returnable. View return policy
Catalog No. NC2232903 Supplier Alpha Resources Inc Supplier No. AR642
Certified Reference Material (CRM), Titanium pin, Hydrogen 0.0046%, Oxygen 0.0453%, Nitrogen 0.0025%, Nominal Weight 0.1 g per pin, Minimum Sample Size 1 pin