Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydridotetrakis(triphenylphosphine)rhodium(I), TCI America™
SDP

Catalog No. H1317200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1317200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1317200MG Supplier TCI America Supplier No. H1317200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18284-36-1
Molecular Formula C72H61P4Rh
Molecular Weight (g/mol) 1153.08
MDL Number MFCD00015867
InChI Key QTTJMGBPWXLZKV-UHFFFAOYSA-N
Synonym hrh pph3 4, hydridotetrakis triphenylphosphine rhodium i, tetrakis triphenylphosphine rhodium hydride, tetrakis triphenylphosphine rhodium, tetrakis triphenylphosphine rhodium i hydride, molecular weight:456.19, molecular formula:c16h26o2rh2, cas number:73468-85-6, rhodium, tetrakis triphenylphosphine, hydridotetrakis triphenylphosphine rhodium
PubChem CID 11981874
IUPAC Name tetrakis(triphenylphosphane) λ¹-rhodium
SMILES [RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 146°C
Quantity 200 mg
Formula Weight 1153.08
Physical Form Crystalline Powder
Chemical Name or Material Hydridotetrakis(triphenylphosphine)rhodium(I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.