Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrindantin Dihydrate 97.0+%, TCI America™
SDP

Catalog No. H01815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H01815G Supplier TCI America Supplier No. H01815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16289-95-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149242
InChI Key QIAIXVRAKYAOGJ-UHFFFAOYSA-N
PubChem CID 23216254
IUPAC Name 2,2'-dihydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1',3,3'-tetrone dihydrate
SMILES O.O.OC1(C(=O)C2=CC=CC=C2C1=O)C1(O)C(=O)C2=CC=CC=C2C1=O

Specifications

Quantity 5 g
Formula Weight 322.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hydrindantin Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.