Learn More
Medchemexpress LLC Hydrindantin hydrate | 5950-69-6 | MFCD00149242 | 98.0% | 358.30 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW011112
Encompass_Preferred
Hydrindantin hydrate is a biochemical assay reagent utilized for the quantitative determination of amino acids.
  • Biochemical assay reagent
  • Used for quantitation determination of amino acid
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-53334