Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Hydrion™ Jumbo Insta-Chek Display pH Papers

Catalog No. 14850125 Shop All Fisherbrand Products
Change view
Click to view available options
pH Range:
0 to 13
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14-850-125 0 to 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-850-125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General purpose pH papers with distinct color match for ranges 0 to 13.

  • Jumbo size pH papers
  • Large color charts
  • Enables clearer, quicker color matching
  • Packaged in plastic dispenser
  • pH range: 0 to 13
  • Dimensions: 1.27 x 1524cm (0.5 x 600 in.)
Note: Image above is representative only. Actual product may vary in appearance.

Specifications

Color Indicator All Ranges
pH Range 0 to 13
Format Roll
For Use With (Application) Determination of pH
Includes Dispenser
Type pH Paper, Jumbo

For diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.