Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrohm USA 0.1 N Hydrochloric Acid, 6 bottles, 1 L each
SDP

Supplier:  Metrohm USA LABTI90051

Encompass

Metrohm USA's 0.1 N Hydrochloric Acid is a high-quality reagent packaged in a case of six 1-liter bottles suitable for various scientific applications. This reliable chemical ensures accuracy for precise analytical procedures like titrations and pH adjustments. Its packaging minimizes waste and enhances ease of handling, making it ideal for ongoing laboratory operations.

  • Reliable chemical reagent for lab needs
  • Exact 0.1 N concentration for analytical accuracy
  • Convenient 1 L bottles reduce waste
  • Packaged in a case of six bottles for ample supply
  • Suitable for titration, pH adjustment, and chemical analysis

Catalog No. NC0387458


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.