Learn More
Dexsil Corporation HYDRO CHLOR Q TEST KIT 12/PK
Catalog No. NC0175845
Encompass
Field Test Kit for Organic Chlorine Determination for Oil/Water Mixture and Used Antifreeze/Coolant between 200-4000ppm
Catalog No. NC0175845 Supplier Dexsil Corporation Supplier No. HYDROSP
Field Test Kit for Organic Chlorine Determination for Oil/Water Mixture and Used Antifreeze/Coolant between 200-4000ppm