Learn More
MCR Safety Hydroblast™ 3 Goggle
Catalog No. 19812775 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-812-775 Gray
19-812-774 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-812-775 Supplier MCR Safety Supplier No. HB1322PF
This item is not returnable. View return policy
MCR Safety Hydroblast™ 3 Goggle features a unique co-injected super soft TPR seal that allows for better, yet more comfortable fit
  • Low profile athletic design
  • 7-Point Ratchet action temple for all day comfort and adjustability
  • Dielectric
  • Appropriate for use in chemical environments
Specifications
Lens Coating Scratch-resistant
Lens Tint Gray
Frame Color Blue
Ventilation Details Indirect vent