Learn More
Mac-Mod HydroBond AQ C8, 100 A, 3 um, 2.1 x 50 mm HPLC Column
SDP
Catalog No. 5090002162
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-900-02162 Supplier Mac-Mod Supplier No. MMHB821053
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.