Learn More
Strem, An Ascensus Company CAS# 10035-10-6. 250g. Hydrobromic acid, 48%. MFCD00011323
SDP
Catalog No. 5081532
Encompass
CAS# 10035-10-6. 250g. Hydrobromic acid, 48%. MFCD00011323. Molecular Weight: 80.92. Molecular Formula: HBr. Color/form: colorless liq. Strem# 93-3504. http://www.strem.com/catalog/v/93-3504/
Catalog No. 50-815-32 Supplier Strem, An Ascensus Company Supplier No. 933504250G
CAS# 10035-10-6. 250g. Hydrobromic acid, 48%. MFCD00011323. Molecular Weight: 80.92. Molecular Formula: HBr. Color/form: colorless liq. Strem# 93-3504. http://www.strem.com/catalog/v/93-3504/