Learn More
Stoelting™ Schwartz T-Clip
SDP
Catalog No. 10001260 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Slight angle
Straight, Smooth
Strong angle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-260 Slight angle
10-001-259 Straight, Smooth
10-001-261 Strong angle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-001-260 Supplier Stoelting™ Supplier No. 5210675
Add to Cart
Add to Cart
Stoelting™ Schwartz T-Clips provide a very easy and secure method of stopping blood flow while performing procedures on vessels temporarily.
  • Using the applying forceps, 52106-73, surgical clips can easily be position on the vessels. The locking forceps allows for easy, precise, and secure grasping of the micro surgical clips. Simply grasp the clip with the forceps to open the jaw, slid it over the vessels and release.
  • To remove surgical clips, simply perform the procedure in reverse.
  • This procedure does not compromise the vessels integrity.
Specifications
For Use With (Application) Neuroscience
Size 1 in.
Model Slight angle