Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrobromic acid solution
Catalog No. 502900623
Encompass
Hydrobromic acid SOLUTIONon (HBr) is a clear yellow or brown colored liquid. Its reaction with K has been studied by a molecular beam technique.
Catalog No. 50-290-0623 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1873550ML
Hydrobromic acid SOLUTIONon (HBr) is a clear yellow or brown colored liquid. Its reaction with K has been studied by a molecular beam technique.